Assay Details
Gene Symbol: LOC101697420 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021240765.1 | XP_021096424.1 | 6 - 7 | 655 | 84 | |
XM_021240766.1 | XP_021096425.1 | 6 - 7 | 633 | 84 | ||
XM_021240767.1 | XP_021096426.1 | 6 - 7 | 708 | 84 | ||
XM_021240768.1 | XP_021096427.1 | 6 - 7 | 677 | 84 | ||
XM_021240769.1 | XP_021096428.1 | 5 - 6 | 532 | 84 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
