Assay Details
| Gene Symbol: CUNH4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247477.1 | XP_021103136.1 | 1 - 2 | 80 | 79 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

CUNH4orf45| Gene Symbol: CUNH4orf45 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021247477.1 | XP_021103136.1 | 1 - 2 | 80 | 79 | |