Assay Details
Gene Symbol: Gorasp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004835235.3 | XP_004835292.1 | 4 - 5 | 517 | 60 | |
XM_021259792.1 | XP_021115451.1 | 4 - 5 | 517 | 60 | ||
XM_021259798.1 | XP_021115457.1 | 4 - 5 | 2296 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() |
