Assay Details
Gene Symbol: Trim56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004840154.2 | XP_004840211.1 | 2 - 3 | 297 | 108 | |
XM_021254476.1 | XP_021110135.1 | 2 - 3 | 331 | 108 | ||
XM_021254477.1 | XP_021110136.1 | 2 - 3 | 326 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004840154.2, XM_021254476.1, XM_021254477.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
