Assay Details
| Gene Symbol: LOC101707236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853447.2 | XP_004853504.2 | 6 - 7 | 1216 | 83 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101707236| Gene Symbol: LOC101707236 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004853447.2 | XP_004853504.2 | 6 - 7 | 1216 | 83 | |
Set Membership: |
|