Assay Details
| Gene Symbol: Gfap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859188.3 | XP_004859245.1 | 4 - 5 | 958 | 93 | |
| XM_004859189.3 | XP_004859246.1 | 4 - 5 | 958 | 93 | ||
| XM_021240220.1 | XP_021095879.1 | 4 - 5 | 958 | 93 | ||
| XM_021240221.1 | XP_021095880.1 | 4 - 5 | 958 | 93 | ||
More Information
Set Membership: |
|

Gfap
Genomic Map