Assay Details
| Gene Symbol: Dmwd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867127.3 | XP_004867184.1 | 4 - 5 | 1997 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dmwd| Gene Symbol: Dmwd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004867127.3 | XP_004867184.1 | 4 - 5 | 1997 | 88 | |
Set Membership: |
|