Assay Details
| Gene Symbol: LOC101708308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073922.2 | XP_012929376.1 | 2 - 3 | 145 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101708308| Gene Symbol: LOC101708308 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013073922.2 | XP_012929376.1 | 2 - 3 | 145 | 88 | |
Set Membership: |
|