Assay Details
Gene Symbol: Znf793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013077442.2 | XP_012932896.1 | 6 - 7 | 867 | 81 | |
XM_021239511.1 | XP_021095170.1 | 6 - 7 | 859 | 81 | ||
XM_021239513.1 | XP_021095172.1 | 5 - 6 | 766 | 81 | ||
XM_021239514.1 | XP_021095173.1 | 7 - 8 | 960 | 81 | ||
XM_021239515.1 | XP_021095174.1 | 5 - 6 | 1274 | 81 | ||
XM_021239516.1 | XP_021095175.1 | 6 - 7 | 897 | 81 | ||
XM_021239517.1 | XP_021095176.1 | 6 - 7 | 859 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
