Assay Details
Gene Symbol: Tmem217 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021259902.1 | XP_021115561.1 | 2 - 3 | 494 | 74 | |
XM_021259903.1 | XP_021115562.1 | 3 - 4 | 224 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
