Assay Details
| Gene Symbol: Pdzrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004854215.3 | XP_004854272.1 | 7 - 8 | 1868 | 60 | |
| XM_013075654.2 | XP_012931108.1 | 5 - 6 | 708 | 60 | ||
| XM_013075656.2 | XP_012931110.1 | 5 - 6 | 967 | 60 | ||
More Information
Set Membership: |
|

Pdzrn3
Genomic Map