Assay Details
Gene Symbol: Ecm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870079.3 | XP_004870136.1 | 5 - 6 | 1348 | 72 | |
XM_013067568.2 | XP_012923022.1 | 5 - 6 | 1184 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
