Assay Details
Gene Symbol: LOC101697194 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013070401.2 | XP_012925855.1 | 3 - 4 | 418 | 104 | |
XM_021254595.1 | XP_021110254.1 | 3 - 4 | 418 | 104 | ||
XM_021254596.1 | XP_021110255.1 | 3 - 4 | 418 | 104 | ||
XM_021254597.1 | XP_021110256.1 | 3 - 4 | 418 | 104 | ||
XM_021254598.1 | XP_021110257.1 | 3 - 4 | 418 | 104 | ||
XM_021254599.1 | XP_021110258.1 | 3 - 4 | 418 | 104 | ||
XM_021254600.1 | XP_021110259.1 | 3 - 4 | 418 | 104 | ||
XM_021254601.1 | XP_021110260.1 | 3 - 4 | 418 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
