Assay Details
| Gene Symbol: LOC101696949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071579.2 | XP_012927033.1 | 7 - 8 | 1108 | 113 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101696949| Gene Symbol: LOC101696949 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013071579.2 | XP_012927033.1 | 7 - 8 | 1108 | 113 | |
Set Membership: |
|