Assay Details
| Gene Symbol: Shroom4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021249971.1 | XP_021105630.1 | 9 - 10 | 5304 | 61 | |
| XM_021249972.1 | XP_021105631.1 | 8 - 9 | 4472 | 61 | ||
More Information
Set Membership: |
|

Shroom4
Genomic Map