Assay Details
Gene Symbol: Mapk1ip1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004835494.3 | XP_004835551.1 | 3 - 4 | 883 | 98 | |
XM_013074603.2 | XP_012930057.1 | 4 - 5 | 984 | 98 | ||
XM_013074606.2 | XP_012930060.1 | 4 - 5 | 966 | 98 | ||
XM_013074610.2 | XP_012930064.1 | 3 - 4 | 906 | 98 | ||
XM_021261887.1 | XP_021117546.1 | 3 - 4 | 969 | 98 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
