Assay Details
Gene Symbol: Clec7a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845883.2 | XP_004845940.1 | 1 - 2 | 211 | 72 | |
XM_004845884.2 | XP_004845941.1 | 1 - 2 | 211 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() |
