Assay Details
Gene Symbol: Kdm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855987.3 | XP_004856044.1 | 3 - 4 | 889 | 52 | |
XM_004855988.3 | XP_004856045.1 | 4 - 5 | 1036 | 52 | ||
XM_004855989.3 | XP_004856046.1 | 3 - 4 | 892 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() |
