Assay Details
Gene Symbol: Cnot9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864583.2 | XP_004864640.1 | 2 - 3 | 633 | 73 | |
XM_013065471.2 | XP_012920925.1 | 2 - 3 | 633 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
