Assay Details
Gene Symbol: Sdhaf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004874330.2 | XP_004874387.1 | 2 - 3 | 360 | 90 | |
XM_021251879.1 | XP_021107538.1 | 2 - 3 | 385 | 90 | ||
XM_021251881.1 | XP_021107540.1 | 2 - 3 | 408 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() |
