Assay Details
Gene Symbol: Ralyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004841991.3 | XP_004842048.1 | 6 - 7 | 899 | 123 | |
XM_013071200.2 | XP_012926654.1 | 6 - 7 | 815 | 123 | ||
XM_013071202.2 | XP_012926656.1 | 5 - 6 | 781 | 123 | ||
XM_013071204.2 | XP_012926658.1 | 7 - 8 | 867 | 123 | ||
XM_013071205.2 | XP_012926659.1 | 6 - 7 | 883 | 123 | ||
XM_013071206.2 | XP_012926660.1 | 6 - 7 | 629 | 123 | ||
XM_013071208.2 | XP_012926662.1 | 6 - 7 | 822 | 123 | ||
XM_013071209.2 | XP_012926663.1 | 5 - 6 | 477 | 123 | ||
XM_021255980.1 | XP_021111639.1 | 6 - 7 | 1361 | 123 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
