Assay Details
| Gene Symbol: Tmprss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842441.3 | XP_004842498.2 | 6 - 7 | 760 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tmprss3| Gene Symbol: Tmprss3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004842441.3 | XP_004842498.2 | 6 - 7 | 760 | 58 | |
Set Membership: |
|