Assay Details
| Gene Symbol: Lrp12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850272.3 | XP_004850329.1 | 5 - 6 | 1859 | 76 | |
| XM_004850273.3 | XP_004850330.1 | 5 - 6 | 1772 | 76 | ||
| XM_021262803.1 | XP_021118462.1 | 4 - 5 | 1715 | 76 | ||
More Information
Set Membership: |
|

Lrp12
Genomic Map