Assay Details
Gene Symbol: CUNH1orf109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004850990.3 | XP_004851047.1 | 2 - 3 | 601 | 62 | |
XM_013074426.2 | XP_012929880.1 | 2 - 3 | 597 | 62 | ||
XM_021263362.1 | XP_021119021.1 | 2 - 3 | 606 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
