Assay Details
Gene Symbol: Prrx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865244.2 | XP_004865301.1 | 2 - 3 | 549 | 63 | |
XM_021244676.1 | XP_021100335.1 | 2 - 3 | 549 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
