Assay Details
Gene Symbol: Nwd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004867675.3 | XP_004867732.1 | 6 - 7 | 1417 | 71 | |
XM_021246677.1 | XP_021102336.1 | 8 - 9 | 1563 | 71 | ||
XM_021246678.1 | XP_021102337.1 | 7 - 8 | 1280 | 71 | ||
XM_021246679.1 | XP_021102338.1 | 6 - 7 | 1360 | 71 | ||
XM_021246680.1 | XP_021102339.1 | 4 - 5 | 1937 | 71 | ||
XM_021246683.1 | XP_021102342.1 | 4 - 5 | 1047 | 71 | ||
XM_021246684.1 | XP_021102343.1 | 2 - 3 | 3394 | 71 | ||
XM_021246685.1 | XP_021102344.1 | 2 - 3 | 685 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
