Assay Details
Gene Symbol: Ubash3b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004870908.3 | XP_004870965.1 | 5 - 6 | 1342 | 64 | |
XM_004870909.2 | XP_004870966.1 | 7 - 8 | 1316 | 64 | ||
XM_021249040.1 | XP_021104699.1 | 5 - 6 | 1344 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
