Assay Details
| Gene Symbol: Nanos3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872444.3 | XP_004872501.1 | 1 - 2 | 2474 | 69 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nanos3| Gene Symbol: Nanos3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004872444.3 | XP_004872501.1 | 1 - 2 | 2474 | 69 | |
Set Membership: |
|