Assay Details
Gene Symbol: LOC101710340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068106.2 | XP_012923560.1 | 10 - 11 | 1032 | 97 | |
XM_021249801.1 | XP_021105460.1 | 10 - 11 | 1032 | 97 | ||
XM_021249802.1 | XP_021105461.1 | 10 - 11 | 1033 | 97 |
