Assay Details
| Gene Symbol: LOC101710340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068106.2 | XP_012923560.1 | 21 - 22 | 2490 | 87 | |
| XM_021249801.1 | XP_021105460.1 | 21 - 22 | 2490 | 87 | ||
| XM_021249802.1 | XP_021105461.1 | 21 - 22 | 2491 | 87 | ||

LOC101710340
Genomic Map