Assay Details
| Gene Symbol: LOC101710340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068106.2 | XP_012923560.1 | 24 - 25 | 2801 | 109 | |
| XM_021249801.1 | XP_021105460.1 | 24 - 25 | 2801 | 109 | ||
| XM_021249802.1 | XP_021105461.1 | 24 - 25 | 2802 | 109 | ||
More Information
Set Membership: |
|

LOC101710340
Genomic Map