Assay Details
Gene Symbol: LOC101710340 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068106.2 | XP_012923560.1 | 2 - 3 | 224 | 61 | |
XM_021249801.1 | XP_021105460.1 | 2 - 3 | 224 | 61 | ||
XM_021249802.1 | XP_021105461.1 | 2 - 3 | 225 | 61 |

More Information
Set Membership: |
![]() ![]() |
