Assay Details
| Gene Symbol: LOC101710340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068106.2 | XP_012923560.1 | 30 - 31 | 3571 | 83 | |
| XM_021249801.1 | XP_021105460.1 | 29 - 30 | 3475 | 83 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101710340| Gene Symbol: LOC101710340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068106.2 | XP_012923560.1 | 30 - 31 | 3571 | 83 | |
| XM_021249801.1 | XP_021105460.1 | 29 - 30 | 3475 | 83 | ||