Assay Details
| Gene Symbol: LOC101710340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068106.2 | XP_012923560.1 | 33 - 34 | 3951 | 87 | |
| XM_021249801.1 | XP_021105460.1 | 32 - 33 | 3855 | 87 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101710340| Gene Symbol: LOC101710340 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013068106.2 | XP_012923560.1 | 33 - 34 | 3951 | 87 | |
| XM_021249801.1 | XP_021105460.1 | 32 - 33 | 3855 | 87 | ||