Assay Details
| Gene Symbol: Dglucy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248244.1 | XP_021103903.1 | 4 - 5 | 513 | 96 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dglucy| Gene Symbol: Dglucy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248244.1 | XP_021103903.1 | 4 - 5 | 513 | 96 | |
Set Membership: |
|