Assay Details
Gene Symbol: Lrrc74a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837490.2 | XP_004837547.2 | 4 - 5 | 769 | 67 | |
XM_021246041.1 | XP_021101700.1 | 5 - 6 | 893 | 67 | ||
XM_021246047.1 | XP_021101706.1 | 5 - 6 | 893 | 67 | ||
XM_021246055.1 | XP_021101714.1 | 6 - 7 | 802 | 67 | ||
XM_021246058.1 | XP_021101717.1 | 5 - 6 | 887 | 67 | ||
XM_021246061.1 | XP_021101720.1 | 6 - 7 | 794 | 67 | ||
XM_021246067.1 | XP_021101726.1 | 4 - 5 | 775 | 67 | ||
XM_021246075.1 | XP_021101734.1 | 4 - 5 | 769 | 67 | ||
XM_021246082.1 | XP_021101741.1 | 5 - 6 | 893 | 67 | ||
XM_021246092.1 | XP_021101751.1 | 5 - 6 | 615 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() |
