Assay Details
Gene Symbol: Sdsl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004843735.3 | XP_004843792.3 | 4 - 5 | 799 | 56 | |
XM_021257404.1 | XP_021113063.1 | 4 - 5 | 790 | 56 | ||
XM_021257405.1 | XP_021113064.1 | 8 - 9 | 1653 | 56 | ||
XM_021257406.1 | XP_021113065.1 | 7 - 8 | 1411 | 56 | ||
XM_021257407.1 | XP_021113066.1 | 7 - 8 | 1194 | 56 | ||
XM_021257408.1 | XP_021113067.1 | 5 - 6 | 637 | 56 | ||
XM_021257409.1 | XP_021113068.1 | 7 - 8 | 1443 | 56 | ||
XM_021257410.1 | XP_021113069.1 | 5 - 6 | 690 | 56 | ||
XM_021257411.1 | XP_021113070.1 | 5 - 6 | 624 | 56 | ||
XM_021257412.1 | XP_021113071.1 | 8 - 9 | 1616 | 56 | ||
XM_021257413.1 | XP_021113072.1 | 7 - 8 | 1374 | 56 | ||
XM_021257414.1 | XP_021113073.1 | 8 - 9 | 1745 | 56 | ||
XM_021257415.1 | XP_021113074.1 | 7 - 8 | 1157 | 56 | ||
XM_021257416.1 | XP_021113075.1 | 5 - 6 | 600 | 56 | ||
XM_021257417.1 | XP_021113076.1 | 5 - 6 | 653 | 56 | ||
XM_021257418.1 | XP_021113077.1 | 5 - 6 | 587 | 56 | ||
XM_021257419.1 | XP_021113078.1 | 4 - 5 | 500 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
