Assay Details
Gene Symbol: Crb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865621.3 | XP_004865678.2 | 6 - 7 | 800 | 102 | |
XM_004865622.3 | XP_004865679.2 | 6 - 7 | 791 | 102 | ||
XM_004865623.3 | XP_004865680.2 | 5 - 6 | 743 | 102 | ||
XM_004865624.3 | XP_004865681.1 | 4 - 5 | 416 | 102 | ||
XM_021245135.1 | XP_021100794.1 | 2 - 3 | 284 | 102 | ||
XM_021245136.1 | XP_021100795.1 | 5 - 6 | 562 | 102 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
