Assay Details
Gene Symbol: Kcnn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871336.2 | XP_004871393.1 | 2 - 3 | 1252 | 63 | |
XM_004871337.3 | XP_004871394.1 | 2 - 3 | 986 | 63 | ||
XM_004871338.2 | XP_004871395.1 | 2 - 3 | 986 | 63 | ||
XM_021249353.1 | XP_021105012.1 | 2 - 3 | 538 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
