Assay Details
Gene Symbol: Ccdc24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871917.3 | XP_004871974.1 | 4 - 5 | 579 | 80 | |
XM_013068164.2 | XP_012923618.1 | 4 - 5 | 579 | 80 | ||
XM_021249951.1 | XP_021105610.1 | 4 - 5 | 593 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
