Assay Details
Gene Symbol: Aurkaip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013065071.2 | XP_012920525.1 | 3 - 4 | 606 | 61 | |
XM_021243592.1 | XP_021099251.1 | 2 - 3 | 907 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
