Assay Details
Gene Symbol: Fez2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013070170.2 | XP_012925624.1 | 4 - 5 | 667 | 70 | |
XM_013070171.2 | XP_012925625.1 | 4 - 5 | 665 | 70 | ||
XM_021253716.1 | XP_021109375.1 | 4 - 5 | 669 | 70 | ||
XM_021253717.1 | XP_021109376.1 | 4 - 5 | 667 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
