Assay Details
Gene Symbol: Stim2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013073005.2 | XP_012928459.1 | 11 - 12 | 2211 | 71 | |
XM_021260206.1 | XP_021115865.1 | 11 - 12 | 1835 | 71 | ||
XM_021260207.1 | XP_021115866.1 | 11 - 12 | 1638 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
