Assay Details
| Gene Symbol: LOC101713044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840458.2 | XP_004840515.1 | 12 - 13 | 2387 | 104 | |
| XM_021254747.1 | XP_021110406.1 | 12 - 13 | 2390 | 104 | ||
| XM_021254748.1 | XP_021110407.1 | 12 - 13 | 2354 | 104 | ||
| XM_021254750.1 | XP_021110409.1 | 12 - 13 | 2361 | 104 | ||
| XM_021254751.1 | XP_021110410.1 | 12 - 13 | 2336 | 104 | ||
| XM_021254752.1 | XP_021110411.1 | 12 - 13 | 2393 | 104 | ||
| XM_021254753.1 | XP_021110412.1 | 12 - 13 | 2368 | 104 | ||
| XM_021254754.1 | XP_021110413.1 | 11 - 12 | 2159 | 104 | ||
| XM_021254755.1 | XP_021110414.1 | 12 - 13 | 2389 | 104 | ||
| XM_021254756.1 | XP_021110415.1 | 9 - 10 | 2722 | 104 | ||
| XM_021254757.1 | XP_021110416.1 | 8 - 9 | 1415 | 104 | ||
| XM_021254758.1 | XP_021110417.1 | 12 - 13 | 2335 | 104 | ||
More Information
Set Membership: |
|

LOC101713044
Genomic Map