Assay Details
| Gene Symbol: LOC101713044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840458.2 | XP_004840515.1 | 16 - 17 | 2870 | 87 | |
| XM_021254747.1 | XP_021110406.1 | 16 - 17 | 2873 | 87 | ||
| XM_021254748.1 | XP_021110407.1 | 16 - 17 | 2837 | 87 | ||
| XM_021254750.1 | XP_021110409.1 | 16 - 17 | 2844 | 87 | ||
| XM_021254751.1 | XP_021110410.1 | 16 - 17 | 2819 | 87 | ||
| XM_021254753.1 | XP_021110412.1 | 16 - 17 | 2851 | 87 | ||
| XM_021254754.1 | XP_021110413.1 | 15 - 16 | 2642 | 87 | ||
| XM_021254755.1 | XP_021110414.1 | 16 - 17 | 2872 | 87 | ||
| XM_021254756.1 | XP_021110415.1 | 13 - 14 | 3205 | 87 | ||
| XM_021254757.1 | XP_021110416.1 | 12 - 13 | 1898 | 87 | ||
| XM_021254758.1 | XP_021110417.1 | 16 - 17 | 2818 | 87 | ||

LOC101713044
Genomic Map