Assay Details
| Gene Symbol: LOC101713044 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840458.2 | XP_004840515.1 | 7 - 8 | 1574 | 52 | |
| XM_021254747.1 | XP_021110406.1 | 7 - 8 | 1577 | 52 | ||
| XM_021254748.1 | XP_021110407.1 | 7 - 8 | 1541 | 52 | ||
| XM_021254750.1 | XP_021110409.1 | 7 - 8 | 1548 | 52 | ||
| XM_021254752.1 | XP_021110411.1 | 7 - 8 | 1580 | 52 | ||
| XM_021254753.1 | XP_021110412.1 | 7 - 8 | 1555 | 52 | ||
| XM_021254755.1 | XP_021110414.1 | 7 - 8 | 1576 | 52 | ||
| XM_021254756.1 | XP_021110415.1 | 4 - 5 | 1909 | 52 | ||
| XM_021254757.1 | XP_021110416.1 | 3 - 4 | 602 | 52 | ||
More Information
Set Membership: |
|

LOC101713044
Genomic Map