Assay Details
| Gene Symbol: LOC101717384 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840608.3 | XP_004840665.1 | 1 - 2 | 143 | 100 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004840608.3 transcripts.
Set Membership: |
|

LOC101717384
Genomic Map