Assay Details
| Gene Symbol: Fam216a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004843820.3 | XP_004843877.1 | 4 - 5 | 536 | 59 | |
| XM_004843821.3 | XP_004843878.1 | 4 - 5 | 529 | 59 | ||
| XM_004843823.3 | XP_004843880.1 | 4 - 5 | 472 | 59 | ||
| XM_004843824.3 | XP_004843881.1 | 4 - 5 | 465 | 59 | ||
More Information
Set Membership: |
|

Fam216a
Genomic Map