Assay Details
| Gene Symbol: Rpia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844546.2 | XP_004844603.1 | 1 - 2 | 322 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Rpia| Gene Symbol: Rpia | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004844546.2 | XP_004844603.1 | 1 - 2 | 322 | 58 | |
Set Membership: |
|