Assay Details
Gene Symbol: LOC101715566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848494.3 | XP_004848551.1 | 3 - 4 | 682 | 107 | |
XM_021261384.1 | XP_021117043.1 | 8 - 9 | 1433 | 107 | ||
XM_021261385.1 | XP_021117044.1 | 9 - 10 | 1228 | 107 | ||
XM_021261386.1 | XP_021117045.1 | 8 - 9 | 1298 | 107 | ||
XM_021261387.1 | XP_021117046.1 | 4 - 5 | 561 | 107 | ||
XM_021261388.1 | XP_021117047.1 | 3 - 4 | 452 | 107 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
